missing translation for 'onlineSavingsMsg'
Learn More

3'-Fluoro-4'-methoxyacetophenone, 99%

Catalog No. AAA1897206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1897206 5 g
AAA1897214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1897206 Supplier Thermo Scientific Chemicals Supplier No. A1897206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 455-91-4 | C9H9FO2 | 168.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 455-91-4
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD00026219
InChI Key LQASUDVYVOFKNK-UHFFFAOYSA-N
Synonym 3'-fluoro-4'-methoxyacetophenone, 3-fluoro-4-methoxyacetophenone, 1-3-fluoro-4-methoxyphenyl ethanone, 1-3-fluoro-4-methoxyphenyl ethan-1-one, ethanone, 1-3-fluoro-4-methoxyphenyl, 3'-fluoro-4-methoxyacetophenone, acetophenone, 3'-fluoro-4'-methoxy, 1-3-fluoro-4-methoxy-phenyl-ethanone, 1-3-fluoro-4-methoxyphenyl-1-ethanone, 1-3-fluoro-4-methoxy-phenyl ethanone
PubChem CID 96828
IUPAC Name 1-(3-fluoro-4-methoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)OC)F

Specifications

Melting Point 90°C to 94°C
Boiling Point 147°C to 148°C (20 mmHg)
Quantity 5 g
Beilstein 2084062
Formula Weight 168.17
Percent Purity 99%
Chemical Name or Material 3'-Fluoro-4'-methoxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-253-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only