missing translation for 'onlineSavingsMsg'
Learn More

3'-Fluoro-4'-hydroxyacetophenone 97.0+%, TCI America™

Catalog No. F08435G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08435G 5 g
F08431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08435G Supplier TCI America Supplier No. F08435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 403-14-5
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00215840
InChI Key GSWTXZXGONEVJC-UHFFFAOYSA-N
Synonym 3'-fluoro-4'-hydroxyacetophenone, 1-3-fluoro-4-hydroxyphenyl ethanone, 1-3-fluoro-4-hydroxyphenyl-1-ethanone, 1-3-fluoro-4-hydroxyphenyl ethan-1-one, 3-fluoro-4-hydroxyacetophenone, ethanone, 1-3-fluoro-4-hydroxyphenyl, 1-3-fluoro-4-hydroxy-phenyl-ethanone, 1-acetyl-3-fluoro-4-hydroxybenzene, pubchem3492, acmc-20a0dy
PubChem CID 2737325
IUPAC Name 1-(3-fluoro-4-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)O)F

Specifications

Melting Point 127°C
Quantity 5 g
Formula Weight 154.14
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Fluoro-4′-hydroxyacetophenone