missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(4-morpholinyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H59970MD

 View more versions of this product

Catalog No. AAH59970MD


Only null left
Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Boronate esters are air- and chromatography-stable and suitable for spectroscopic study. The Suzuki-Miyaura cross-coupling reaction can be used with boronate esters.

Notes
Store at room temperature. Keep away from strong oxidizing agents.
Specifications

Specifications

3-Fluoro-4-(4-morpholinyl)benzeneboronic acid pinacol ester
C16H23BFNO3
250 mg
YOJYRVSDQHWBGS-UHFFFAOYSA-N
4-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
70975109
95%
873431-46-0
MFCD22988989
4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 3-fluoro-4-4-morpholinyl benzeneboronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)N3CCOCC3)F
307.172
307.17

Chemical Identifiers

873431-46-0
307.172
YOJYRVSDQHWBGS-UHFFFAOYSA-N
70975109
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)N3CCOCC3)F
C16H23BFNO3
MFCD22988989
4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 3-fluoro-4-4-morpholinyl benzeneboronic acid pinacol ester
4-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions