Learn More
3-Fluoro-2-nitropyridine, 96%
CAS: 54231-35-5 | C5H3FN2O2 | 142.089 g/mol
$294.81 - $294.81
Chemical Identifiers
| CAS | 54231-35-5 |
|---|---|
| Molecular Formula | C5H3FN2O2 |
| Molecular Weight (g/mol) | 142.089 |
| MDL Number | MFCD04114127 |
| InChI Key | IJVFHCSUEBAAOZ-UHFFFAOYSA-N |
| Synonym | 2-nitro-3-fluoropyridine, 3-fluoro-2-nitro-pyridine, pyridine, 3-fluoro-2-nitro, pubchem2185, 3-fluor-2-nitropyridin, pyfn51, 3-fluoro 2-nitro pyridine, pyridine,3-fluoro-2-nitro, pyridine, 3-fluoro-2-nitro-9ci |
| PubChem CID | 2762802 |
| IUPAC Name | 3-fluoro-2-nitropyridine |
| SMILES | C1=CC(=C(N=C1)[N+](=O)[O-])F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3381303
|
Thermo Scientific Chemicals
H3381303 |
1 g |
Each for $294.81
|
|
|||||
Description
A specialty product for proteomics research applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA specialty product for proteomics research applications.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
| 54231-35-5 | |
| 142.089 | |
| IJVFHCSUEBAAOZ-UHFFFAOYSA-N | |
| 2762802 | |
| C1=CC(=C(N=C1)[N+](=O)[O-])F |
| C5H3FN2O2 | |
| MFCD04114127 | |
| 2-nitro-3-fluoropyridine, 3-fluoro-2-nitro-pyridine, pyridine, 3-fluoro-2-nitro, pubchem2185, 3-fluor-2-nitropyridin, pyfn51, 3-fluoro 2-nitro pyridine, pyridine,3-fluoro-2-nitro, pyridine, 3-fluoro-2-nitro-9ci | |
| 3-fluoro-2-nitropyridine |
Specifications
| 54231-35-5 | |
| C5H3FN2O2 | |
| 1 g | |
| Slightly soluble in water. | |
| C1=CC(=C(N=C1)[N+](=O)[O-])F | |
| 142.089 | |
| 142.09 | |
| 3-Fluoro-2-nitropyridine |
| 30°C to 35°C | |
| MFCD04114127 | |
| 2-nitro-3-fluoropyridine, 3-fluoro-2-nitro-pyridine, pyridine, 3-fluoro-2-nitro, pubchem2185, 3-fluor-2-nitropyridin, pyfn51, 3-fluoro 2-nitro pyridine, pyridine,3-fluoro-2-nitro, pyridine, 3-fluoro-2-nitro-9ci | |
| IJVFHCSUEBAAOZ-UHFFFAOYSA-N | |
| 3-fluoro-2-nitropyridine | |
| 2762802 | |
| 96% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only