missing translation for 'onlineSavingsMsg'
Learn More

3'-Fluoro-2'-hydroxyacetophenone, 98%

Catalog No. AAH32854MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32854MD 250 mg
AAH3285403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32854MD Supplier Thermo Scientific Chemicals Supplier No. H32854MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 699-92-3 | C8H7FO2 | 154.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 699-92-3
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD09055140
InChI Key WXTHZWWWCICGAN-UHFFFAOYSA-N
Synonym 3'-fluoro-2'-hydroxyacetophenone, 1-3-fluoro-2-hydroxyphenyl ethanone, 1-3-fluoro-2-hydroxy-phenyl-ethanone, 1-3-fluoro-2-hydroxyphenyl ethan-1-one, 3-fluoro-2-hydroxyacetophenone, ethanone, 1-3-fluoro-2-hydroxyphenyl, 1-3-fluoro-2-hydroxyphenyl-ethanone, pubchem19786, 3;-fluoro-2;-hydroxyacetophenone
PubChem CID 10773176
IUPAC Name 1-(3-fluoro-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(O)C(F)=CC=C1

Specifications

Quantity 250 mg
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material 3'-Fluoro-2'-hydroxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only