missing translation for 'onlineSavingsMsg'
Learn More

3-Ethynylpyridine 98.0+%, TCI America™

Catalog No. E05605G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05605G 5 g
E05601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05605G Supplier TCI America Supplier No. E05605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2510-23-8
Molecular Formula C7H5N
Molecular Weight (g/mol) 103.124
MDL Number MFCD02177459
InChI Key CLRPXACRDTXENY-UHFFFAOYSA-N
Synonym pyridine, 3-ethynyl, 3-pyridylacetylene, 3-ethynyl-pyridine, pyridine, 3-ethynyl-6ci, 7ci, 8ci, 9ci, 3-ethylnylpyridine, 3-ethynyl pyridine, meta-ethynylpyridine, 3-ethylnyl pyridine, 3-pyridyl acetylene
PubChem CID 186003
IUPAC Name 3-ethynylpyridine
SMILES C#CC1=CN=CC=C1

Specifications

Melting Point 36°C
Quantity 5 g
UN Number 1325
Formula Weight 103.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethynylpyridine
TSCA No
Recommended Storage Freezer