missing translation for 'onlineSavingsMsg'
Learn More

3-Ethynylpyridine, 96%

Catalog No. p-3759046
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398070010 1 g Glass bottle
AC398070050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC398070010 Supplier Thermo Scientific Chemicals Supplier No. 398070010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2510-23-8 | C7H5N | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2510-23-8
Molecular Formula C7H5N
Molecular Weight (g/mol) 103.12
MDL Number MFCD02177459
InChI Key CLRPXACRDTXENY-UHFFFAOYSA-N
Synonym pyridine, 3-ethynyl, 3-pyridylacetylene, 3-ethynyl-pyridine, pyridine, 3-ethynyl-6ci, 7ci, 8ci, 9ci, 3-ethylnylpyridine, 3-ethynyl pyridine, meta-ethynylpyridine, 3-ethylnyl pyridine, 3-pyridyl acetylene
PubChem CID 186003
IUPAC Name 3-ethynylpyridine
SMILES C#CC1=CN=CC=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 39°C to 40°C
Color Brown to Colorless
Boiling Point 83°C to 84°C (30.0 mmHg)
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 103.12
Percent Purity 96%
Physical Form Low Melting Solid
Chemical Name or Material 3-Ethynylpyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable solid.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage Product darkens on storage

RUO – Research Use Only