missing translation for 'onlineSavingsMsg'
Learn More

3-Ethynylbenzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Catalog No. p-7034235
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5190203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5190203 Supplier Thermo Scientific Chemicals Supplier No. H5190203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 946168-04-3
Molecular Formula C14H17BO2
Molecular Weight (g/mol) 228.098
MDL Number MFCD16294529
InChI Key SJDFLZSEQGKSDJ-UHFFFAOYSA-N
Synonym 2-3-ethynylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-ethynyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 3-ethynylbenzeneboronic acid pinacol ester, amtb210, 3-ethynylphenyl boronic acid pinacol ester, 2-3-ethynyl-phenyl-4,4,5,5-tetramethyl-1,3,2dioxaborolane
PubChem CID 57416900
IUPAC Name 2-(3-ethynylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C#C

Specifications

Quantity 1 g
Beilstein 11140327
Formula Weight 228.1
Percent Purity 95%
Chemical Name or Material 3-Ethynylbenzeneboronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures