missing translation for 'onlineSavingsMsg'
Learn More

3-Ethynylanisole 97.0+%, TCI America™

Catalog No. E06271G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06271G 1 g
E06275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06271G Supplier TCI America Supplier No. E06271G

Chemical Identifiers

CAS 768-70-7
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00160810
InChI Key ZASXCTCNZKFDTP-UHFFFAOYSA-N
Synonym 3-ethynylanisole, 3-methoxyphenylacetylene, 1-ethynyl-3-methoxy-benzene, benzene, 1-ethynyl-3-methoxy, 3-ethynyl-1-methoxybenzene, 1-etynyl-3-methoxy-benzene, 3-ethynyl-anisole, m-methoxyphenylacetylene, 3-ethynylanisole 96
PubChem CID 640753
IUPAC Name 1-ethynyl-3-methoxybenzene
SMILES COC1=CC=CC(=C1)C#C

Specifications

Color Yellow
Boiling Point 90°C
Quantity 1 g
Formula Weight 132.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Ethynylanisole
TSCA No
Recommended Storage Refrigerator