missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyltoluene, 97%

Catalog No. AAL1878806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1878806 5 g
AAL1878814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1878806 Supplier Thermo Scientific Chemicals Supplier No. L1878806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 620-14-4 | C9H12 | 120.20 g/mol

It is used in the formation of 3-ethylsalicylic acid from 3-Ethyltoluene by Pseudomonas ovalis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the formation of 3-ethylsalicylic acid from 3-Ethyltoluene by Pseudomonas ovalis.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away. Protect from electrostatic charges.

Chemical Identifiers

CAS 620-14-4
Molecular Formula C9H12
Molecular Weight (g/mol) 120.20
MDL Number MFCD00009259
InChI Key ZLCSFXXPPANWQY-UHFFFAOYSA-N
Synonym 3-ethyltoluene, m-ethyltoluene, 1-methyl-3-ethylbenzene, benzene, 1-ethyl-3-methyl, m-ethylmethylbenzene, m-methylethylbenzene, toluene, m-ethyl, 3-methylethylbenzene, 1-ethyl-3-methyl-benzene, unii-737ptd7o7e
PubChem CID 12100
ChEBI CHEBI:77512
IUPAC Name 1-ethyl-3-methylbenzene
SMILES CCC1=CC=CC(C)=C1

Specifications

Melting Point -96°C
Density 0.865
Boiling Point 158°C to 159°C
Flash Point 38°C (100°F)
Refractive Index 1.496
Quantity 5 g
UN Number UN3295
Beilstein 1850821
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 120.2
Percent Purity ≥98%
Chemical Name or Material 3-Ethyltoluene
Show More Show Less
Hazard Category H226-H304-H315-H319-H335
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 210-626-8
RTECSNumber DA0712000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only