missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethylpyridine 98.0+%, TCI America™
$67.11 - $199.46
Chemical Identifiers
| CAS | 536-78-7 |
|---|---|
| Molecular Formula | C7H9N |
| Molecular Weight (g/mol) | 107.156 |
| MDL Number | MFCD00006413 |
| InChI Key | MFEIKQPHQINPRI-UHFFFAOYSA-N |
| Synonym | pyridine, 3-ethyl, 3-ethyl pyridine, beta-lutidine, beta-ethylpyridine, 3-ethyl-pyridine, 5-ethylpyridine, unii-a25i3ez88v, lutidine, beta, 3ethylpyridine, fema no. 3394 |
| PubChem CID | 10823 |
| IUPAC Name | 3-ethylpyridine |
| SMILES | CCC1=CN=CC=C1 |
Chemical Identifiers
| 536-78-7 | |
| 107.156 | |
| MFEIKQPHQINPRI-UHFFFAOYSA-N | |
| 10823 | |
| CCC1=CN=CC=C1 |
| C7H9N | |
| MFCD00006413 | |
| pyridine, 3-ethyl, 3-ethyl pyridine, beta-lutidine, beta-ethylpyridine, 3-ethyl-pyridine, 5-ethylpyridine, unii-a25i3ez88v, lutidine, beta, 3ethylpyridine, fema no. 3394 | |
| 3-ethylpyridine |
Specifications
| 536-78-7 | |
| 165°C | |
| MFCD00006413 | |
| 1993 | |
| MFEIKQPHQINPRI-UHFFFAOYSA-N | |
| 3-ethylpyridine | |
| 10823 | |
| ≥98.0% (GC) | |
| 3-Ethylpyridine |
| Yellow | |
| C7H9N | |
| 5 mL | |
| pyridine, 3-ethyl, 3-ethyl pyridine, beta-lutidine, beta-ethylpyridine, 3-ethyl-pyridine, 5-ethylpyridine, unii-a25i3ez88v, lutidine, beta, 3ethylpyridine, fema no. 3394 | |
| CCC1=CN=CC=C1 | |
| 107.156 | |
| 107.16 | |
| Liquid |
Safety and Handling
EINECSNumber : (9)-0222
TSCA : Yes