missing translation for 'onlineSavingsMsg'
Learn More

3-Ethylpyridine, 98%

Catalog No. AAB2031814
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2031814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2031814 Supplier Thermo Scientific Chemicals Supplier No. B2031814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 536-78-7 | C7H9N | 107.156 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 536-78-7
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00006413
InChI Key MFEIKQPHQINPRI-UHFFFAOYSA-N
Synonym pyridine, 3-ethyl, 3-ethyl pyridine, beta-lutidine, beta-ethylpyridine, 3-ethyl-pyridine, 5-ethylpyridine, unii-a25i3ez88v, lutidine, beta, 3ethylpyridine, fema no. 3394
PubChem CID 10823
IUPAC Name 3-ethylpyridine
SMILES CCC1=CN=CC=C1

Specifications

Melting Point -77°C
Density 0.941
Boiling Point 165°C to 166°C
Flash Point 49°C (120°F)
Odor Unpleasant
Refractive Index 1.502
Quantity 25 g
UN Number UN1993
Beilstein 106479
Merck Index 14,3848
Formula Weight 107.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Ethylpyridine
Show More Show Less
Hazard Category H226-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement

H226-H302-H312-H332-H315-H319-H335

Flammable liquid and vapour.

Harmful if swallowed.

Harmful in contact with skin.

Harmful if inhaled.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 208-647-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only