missing translation for 'onlineSavingsMsg'
Learn More

3-Ethylcyclopentanone, 99%

Catalog No. p-7025100
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2043203 1 g
AAB2043206 5 g
AAB2043214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2043203 Supplier Thermo Scientific Chemicals Supplier No. B2043203

CAS: 10264-55-8 | C7H12O | 112.172 g/mol

3-Ethylcyclopentanone, is used as a chemical reagent, pharmaceutical reagent and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethylcyclopentanone, is used as a chemical reagent, pharmaceutical reagent and pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 10264-55-8
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00045445
InChI Key XERALSLWOPMNRJ-UHFFFAOYSA-N
PubChem CID 139121
IUPAC Name 3-ethylcyclopentan-1-one
SMILES CCC1CCC(=O)C1

Specifications

Boiling Point 171°C
Flash Point 31°C (88°F)
Quantity 1 g
UN Number UN1224
Synonym 3-ethylcyclopentanone, cyclopentanone, 3-ethyl, 3-ethyl cyclopentanone, cyclopentanone,3-ethyl, acmc-20an48, 3-ethyl-cyclopentan-1-one
Solubility Information Soluble in water.
Formula Weight 112.17
Percent Purity 99%
Chemical Name or Material 3-Ethylcyclopentanone
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only