missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethylbenzothiazolium bromide, 98%
CAS: 32446-47-2 | C9H10BrNS | 244.15 g/mol
$127.10 - $418.38
Chemical Identifiers
| CAS | 32446-47-2 |
|---|---|
| Molecular Formula | C9H10BrNS |
| Molecular Weight (g/mol) | 244.15 |
| MDL Number | MFCD00051852 |
| InChI Key | XBWYYTURPBHPFJ-UHFFFAOYSA-M |
| Synonym | 3-ethylbenzothiazolium bromide, benzothiazolium, 3-ethyl-, bromide 1:1, benzothiazolium, 3-ethyl-, bromide, 3-ethyl-1,3-benzothiazol-3-ium bromide, 3-ethylbenzothiazoliumbromide, acmc-209ht4, 3-ethylbenzo d thiazol-3-ium bromide |
| PubChem CID | 182081 |
| IUPAC Name | 3-ethyl-1,3-benzothiazol-3-ium;bromide |
| SMILES | [Br-].CC[N+]1=CSC2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0890309
|
Thermo Scientific Chemicals
L0890309 |
10 g |
N/A
|
|
|||||
|
AAL0890318
|
Thermo Scientific Chemicals
L0890318 |
50 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 32446-47-2 | |
| 244.15 | |
| XBWYYTURPBHPFJ-UHFFFAOYSA-M | |
| 182081 | |
| [Br-].CC[N+]1=CSC2=CC=CC=C12 |
| C9H10BrNS | |
| MFCD00051852 | |
| 3-ethylbenzothiazolium bromide, benzothiazolium, 3-ethyl-, bromide 1:1, benzothiazolium, 3-ethyl-, bromide, 3-ethyl-1,3-benzothiazol-3-ium bromide, 3-ethylbenzothiazoliumbromide, acmc-209ht4, 3-ethylbenzo d thiazol-3-ium bromide | |
| 3-ethyl-1,3-benzothiazol-3-ium;bromide |
Specifications
| 32446-47-2 | |
| C9H10BrNS | |
| 10 g | |
| Hygroscopic | |
| XBWYYTURPBHPFJ-UHFFFAOYSA-M | |
| 3-ethyl-1,3-benzothiazol-3-ium;bromide | |
| 182081 | |
| 98% |
| 205°C to 207°C | |
| MFCD00051852 | |
| 6250086 | |
| 3-ethylbenzothiazolium bromide, benzothiazolium, 3-ethyl-, bromide 1:1, benzothiazolium, 3-ethyl-, bromide, 3-ethyl-1,3-benzothiazol-3-ium bromide, 3-ethylbenzothiazoliumbromide, acmc-209ht4, 3-ethylbenzo d thiazol-3-ium bromide | |
| [Br-].CC[N+]1=CSC2=CC=CC=C12 | |
| 244.15 | |
| 244.16 | |
| 3-Ethylbenzothiazolium bromide |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only