missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethylaniline 98.0+%, TCI America™
$298.25 - $3856.73
Chemical Identifiers
| CAS | 587-02-0 |
|---|---|
| Molecular Formula | C8H11N |
| Molecular Weight (g/mol) | 121.18 |
| MDL Number | MFCD00007818 |
| InChI Key | AMKPQMFZCBTTAT-UHFFFAOYSA-N |
| Synonym | m-ethylaniline, benzenamine, 3-ethyl, 3-ethylbenzenamine, 3-ethylphenylamine, aniline, m-ethyl, unii-w5u3oqy77o, aniline, 3-ethyl, w5u3oqy77o, methylanilin, 3-ethyl aniline |
| PubChem CID | 11475 |
| IUPAC Name | 3-ethylaniline |
| SMILES | CCC1=CC=CC(N)=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E036425ML
|
TCI America
E036425ML |
25 mL |
Each for $298.25
|
|
|||||
|
E0364500ML
|
TCI America
E0364500ML |
500 mL |
Each for $3,856.73
|
|
|||||
Chemical Identifiers
| 587-02-0 | |
| 121.18 | |
| AMKPQMFZCBTTAT-UHFFFAOYSA-N | |
| 11475 | |
| CCC1=CC=CC(N)=C1 |
| C8H11N | |
| MFCD00007818 | |
| m-ethylaniline, benzenamine, 3-ethyl, 3-ethylbenzenamine, 3-ethylphenylamine, aniline, m-ethyl, unii-w5u3oqy77o, aniline, 3-ethyl, w5u3oqy77o, methylanilin, 3-ethyl aniline | |
| 3-ethylaniline |
Specifications
| 587-02-0 | |
| Yellow | |
| C8H11N | |
| 25 mL | |
| m-ethylaniline, benzenamine, 3-ethyl, 3-ethylbenzenamine, 3-ethylphenylamine, aniline, m-ethyl, unii-w5u3oqy77o, aniline, 3-ethyl, w5u3oqy77o, methylanilin, 3-ethyl aniline | |
| CCC1=CC=CC(N)=C1 | |
| 121.18 | |
| 121.18 | |
| Liquid |
| -8°C | |
| 214°C | |
| MFCD00007818 | |
| 2810 | |
| AMKPQMFZCBTTAT-UHFFFAOYSA-N | |
| 3-ethylaniline | |
| 11475 | |
| ≥98.0% (GC,T) | |
| 3-Ethylaniline |
Safety and Handling
EINECSNumber : (3)-0206
RTECSNumber : BX9770000
TSCA : No