missing translation for 'onlineSavingsMsg'
Learn More

3-Ethylaniline 98.0+%, TCI America™

Catalog No. E036425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E036425ML 25 mL
E0364500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E036425ML Supplier TCI America Supplier No. E036425ML

Chemical Identifiers

CAS 587-02-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00007818
InChI Key AMKPQMFZCBTTAT-UHFFFAOYSA-N
Synonym m-ethylaniline, benzenamine, 3-ethyl, 3-ethylbenzenamine, 3-ethylphenylamine, aniline, m-ethyl, unii-w5u3oqy77o, aniline, 3-ethyl, w5u3oqy77o, methylanilin, 3-ethyl aniline
PubChem CID 11475
IUPAC Name 3-ethylaniline
SMILES CCC1=CC=CC(N)=C1

Specifications

Melting Point -8°C
Color Yellow
Boiling Point 214°C
Quantity 25 mL
UN Number 2810
Formula Weight 121.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Ethylaniline
EINECSNumber (3)-0206
RTECSNumber BX9770000
TSCA No