missing translation for 'onlineSavingsMsg'
Learn More

3-(Ethylamino)pyrrolidine 97.0+%, TCI America™

Catalog No. E04315G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E04315G Supplier TCI America Supplier No. E04315G

Chemical Identifiers

CAS 111390-22-8
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00191357
InChI Key OPCPWFHLFKAUEA-UHFFFAOYSA-N
PubChem CID 14280900
IUPAC Name N-ethylpyrrolidin-3-amine
SMILES CCNC1CCNC1

Specifications

Color Colorless
Boiling Point 176°C
Quantity 5 g
UN Number 2734
Formula Weight 114.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Ethylamino)pyrrolidine