missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-5-methylisoxazole-4-carboxylic acid, 97%

Catalog No. p-7030043
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
10 g
Catalog No. Quantity
AAH2757403 1 g
AAH2757409 10 g
2 options

Catalog No. AAH2757403

Supplier: Thermo Scientific Chemicals H2757403

Only null left

CAS: 17147-85-2 | C7H9NO3 | 155.153 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17147-85-2
Molecular Formula C7H9NO3
Molecular Weight (g/mol) 155.153
MDL Number MFCD03422293
InChI Key RSWFHHWVFZWIMV-UHFFFAOYSA-N
Synonym 3-ethyl-5-methylisoxazole-4-carboxylic acid, 3-ethyl-5-methyl-isoxazole-4-carboxylic acid, 3-ethyl-5-methyl-4-isoxazolecarboxylic acid, 4-isoxazolecarboxylic acid, 3-ethyl-5-methyl, acmc-1c8pz, 4-isoxazolecarboxylicacid,3-ethyl-5-methyl, 4-isoxazolecarboxylicacid, 3-ethyl-5-methyl, 4-isoxazolecarboxylic acid, 3-ethyl-5-methyl-8ci,9ci
PubChem CID 2063310
IUPAC Name 3-ethyl-5-methyl-1,2-oxazole-4-carboxylic acid
SMILES CCC1=NOC(=C1C(=O)O)C

Specifications

Melting Point 123°C to 125°C
Quantity 1 g
Formula Weight 155.15
Percent Purity 97%
Chemical Name or Material 3-Ethyl-5-methylisoxazole-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only