missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-5-methylisoxazole-4-carboxylic acid, 97%

Catalog No. AAH2757409
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2757409 10 g
AAH2757403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2757409 Supplier Thermo Scientific Chemicals Supplier No. H2757409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17147-85-2 | C7H9NO3 | 155.153 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17147-85-2
Molecular Formula C7H9NO3
Molecular Weight (g/mol) 155.153
MDL Number MFCD03422293
InChI Key RSWFHHWVFZWIMV-UHFFFAOYSA-N
Synonym 3-ethyl-5-methylisoxazole-4-carboxylic acid, 3-ethyl-5-methyl-isoxazole-4-carboxylic acid, 3-ethyl-5-methyl-4-isoxazolecarboxylic acid, 4-isoxazolecarboxylic acid, 3-ethyl-5-methyl, acmc-1c8pz, 4-isoxazolecarboxylicacid,3-ethyl-5-methyl, 4-isoxazolecarboxylicacid, 3-ethyl-5-methyl, 4-isoxazolecarboxylic acid, 3-ethyl-5-methyl-8ci,9ci
PubChem CID 2063310
IUPAC Name 3-ethyl-5-methyl-1,2-oxazole-4-carboxylic acid
SMILES CCC1=NOC(=C1C(=O)O)C

Specifications

Melting Point 123°C to 125°C
Quantity 10 g
Formula Weight 155.15
Percent Purity 97%
Chemical Name or Material 3-Ethyl-5-methylisoxazole-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only