missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazolium bromide, 98%

Catalog No. p-7039959
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5948506 5 g
AAH5948514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5948506 Supplier Thermo Scientific Chemicals Supplier No. H5948506

CAS: 54016-70-5 | C8H14BrNOS | 252.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54016-70-5
Molecular Formula C8H14BrNOS
Molecular Weight (g/mol) 252.17
MDL Number MFCD00040549
InChI Key BDQRQMLWZJQQKS-UHFFFAOYSA-M
Synonym 3-ethyl-5-2-hydroxyethyl-4-methylthiazolium bromide, 3-ethyl-5-2-hydroxyethyl-4-methylthiazoliumbromide, 3-ethyl-5-2-hydroxyethyl-4-methylthiazol-3-ium bromide, thiazolium, 3-ethyl-5-2-hydroxyethyl-4-methyl-, bromide, 2-3-ethyl-4-methyl-thiazol-3-ium-5-yl ethanol bromide, 3-ethyl-5-2-hydroxyethyl-4-methyl-1,3-thiazol-3-ium bromide, pubchem15236, acmc-1area, ksc272q3h, 3ethyl-5-2-hydroxyethyl-4-methylthiazolium bromide
PubChem CID 9964923
IUPAC Name 2-(3-ethyl-4-methyl-1,3-thiazol-3-ium-5-yl)ethanol;bromide
SMILES CC[N+]1=CSC(=C1C)CCO.[Br-]

Specifications

Melting Point 81°C to 87°C
Quantity 5 g
Beilstein 4165775
Formula Weight 252.17
Percent Purity 98%
Chemical Name or Material 3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazolium bromide
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 258-925-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only