missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-4-methyl-3-pyrrolin-2-one, 98%

Catalog No. p-7036725
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5506503 1 g
AAH5506506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5506503 Supplier Thermo Scientific Chemicals Supplier No. H5506503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 766-36-9 | C7H11NO | 125.17 g/mol

3-Ethyl-4-methyl-3-pyrrolin-2-one is used as an intermediate for the manufacture of medicines. It is used for the preparation of Glimepiride (G410150), and its intermediates and as antidiabetic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethyl-4-methyl-3-pyrrolin-2-one is used as an intermediate for the manufacture of medicines. It is used for the preparation of Glimepiride (G410150), and its intermediates and as antidiabetic agents.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 766-36-9
Molecular Formula C7H11NO
Molecular Weight (g/mol) 125.17
MDL Number MFCD00173861
InChI Key YCTNTSVMJWIYTQ-UHFFFAOYSA-N
Synonym 3-ethyl-4-methyl-3-pyrrolin-2-one, 3-ethyl-4-methyl-3-pyrroline-2-one, 3-ethyl-4-methyl-2,5-dihydro-1h-pyrrol-2-one, 3-ethyl-4-methyl-1h-pyrrol-2 5h-one, 1,5-dihydro-3-ethyl-4-methyl-2h-pyrrol-2-one, 2h-pyrrol-2-one, 3-ethyl-1,5-dihydro-4-methyl, 3-ethyl-4-methyl-2-oxo-3-pyrroline, 3-ethyl-4-methyl-1,5-dihydropyrrol-2-one, 3-ethyl-1,5-dihydro-4-methyl-2h-pyrrol-2-one, pubchem12392
PubChem CID 854146
SMILES CCC1=C(C)CNC1=O

Specifications

Melting Point 100°C to 104°C
Quantity 1 g
Solubility Information Slightly soluble in water.
IUPAC Name 3-ethyl-4-methyl-2,5-dihydro-1H-pyrrol-2-one
Formula Weight 125.17
Percent Purity 98%
Chemical Name or Material 3-Ethyl-4-methyl-3-pyrrolin-2-one
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only