missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-3-pentanol, 97%

Catalog No. AAL0302509
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0302509 10 g
AAL0302518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0302509 Supplier Thermo Scientific Chemicals Supplier No. L0302509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 597-49-9 | C7H16O | 116.20 g/mol

3-Ethyl-3-pentanol was used to study the effect of alkanols on the micropolarity and microviscosity of the head group region, in the reverse micelles of AOT-heptane-water by fluorescence probing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethyl-3-pentanol was used to study the effect of alkanols on the micropolarity and microviscosity of the head group region, in the reverse micelles of AOT-heptane-water by fluorescence probing.

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Store away from acids, acid chlorides and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect against electrostatic charges.

Chemical Identifiers

CAS 597-49-9
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.20
MDL Number MFCD00004484
InChI Key XKIRHOWVQWCYBT-UHFFFAOYSA-N
Synonym 3-ethyl-3-pentanol, triethylcarbinol, 3-pentanol, 3-ethyl, triethylmethanol, triethylmethyl alcohol, tert-heptanol, 3-aethyl-pentanol-3, 3-aethyl-pentanol-3 german, 3-ethyl-3-hydroxypentane, ethyl-3 pentanol-3
PubChem CID 11702
IUPAC Name 3-ethylpentan-3-ol
SMILES CCC(O)(CC)CC

Specifications

Melting Point -13°C
Density 0.846
Boiling Point 143°C
Flash Point 40°C (104°F)
Odor Characteristic
Refractive Index 1.43
Linear Formula (CH3CH2)3COH
Quantity 10 g
UN Number UN1987
Beilstein 1731643
Solubility Information Slightly soluble in water. Soluble in alcohol.
Formula Weight 116.2
Percent Purity 97%
Chemical Name or Material 3-Ethyl-3-pentanol
Show More Show Less
Hazard Category H226
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 209-902-0
RTECSNumber SA6780000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only