missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-3-oxetanemethanol 96.0+%, TCI America™

Catalog No. p-7129059
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E054625G 25 g
E0546100G 100 g
E0546500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E054625G Supplier TCI America Supplier No. E054625G

Chemical Identifiers

CAS 3047-32-3
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00190143
InChI Key UNMJLQGKEDTEKJ-UHFFFAOYSA-N
Synonym 3-ethyl-3-oxetanemethanol, 3-ethyloxetan-3-yl methanol, 3-oxetanemethanol, 3-ethyl, 3-ethyloxetane-3-methanol, unii-622325vbyo, 3-ethyl-3-hydroxymethyloxetane, 3-hydroxymethyl-3-ethyloxetane, 3-ethyl-3-hydroxymethyl oxetane, 3-ethyl-oxetan-3-yl-methanol, 2-hydroxymethyl-2-ethyl-1,3-epoxypropane
PubChem CID 76444
IUPAC Name (3-ethyloxetan-3-yl)methanol
SMILES CCC1(COC1)CO

Specifications

Color Colorless
Boiling Point 110°C
Quantity 25 g
Formula Weight 116.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Ethyl-3-oxetanemethanol
EINECSNumber (5)-6621
TSCA Yes