missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-3-oxetanemethanol, 97%

Catalog No. p-7035770
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5349706 5 g
AAH5349714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5349706 Supplier Thermo Scientific Chemicals Supplier No. H5349706

CAS: 3047-32-3 | C6H12O2 | 116.16 g/mol

3-Ethyl-3-oxetanemethanol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethyl-3-oxetanemethanol is used as pharmaceutical intermediate.

Solubility
Miscible in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 3047-32-3
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00190143
InChI Key UNMJLQGKEDTEKJ-UHFFFAOYSA-N
Synonym 3-ethyl-3-oxetanemethanol, 3-ethyloxetan-3-yl methanol, 3-oxetanemethanol, 3-ethyl, 3-ethyloxetane-3-methanol, unii-622325vbyo, 3-ethyl-3-hydroxymethyloxetane, 3-hydroxymethyl-3-ethyloxetane, 3-ethyl-3-hydroxymethyl oxetane, 3-ethyl-oxetan-3-yl-methanol, 2-hydroxymethyl-2-ethyl-1,3-epoxypropane
PubChem CID 76444
IUPAC Name (3-ethyloxetan-3-yl)methanol
SMILES CCC1(COC1)CO

Specifications

Density 1.019
Boiling Point 96°C (4 mmHg)
Flash Point 108°C (226°F)
Refractive Index 1.453
Quantity 5 g
Solubility Information Miscible in water.
Formula Weight 116.16
Percent Purity 97%
Chemical Name or Material 3-Ethyl-3-oxetanemethanol
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 221-254-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only