missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-3-methylpentane 99.0+%, TCI America™

Catalog No. E02585ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E02585ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E02585ML Supplier TCI America Supplier No. E02585ML
Only null left

Chemical Identifiers

CAS 1067-08-9
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00048630
InChI Key GIEZWIDCIFCQPS-UHFFFAOYSA-N
PubChem CID 14018
IUPAC Name 3-ethyl-3-methylpentane
SMILES CCC(C)(CC)CC

Specifications

Color Colorless
Boiling Point 118°C
Quantity 5 mL
UN Number 1262
Formula Weight 114.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Ethyl-3-methylpentane
EINECSNumber (2)-0008
TSCA No