missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-3-hexanol, 98%, Thermo Scientific Chemicals

Catalog No. AAB2085906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2085906 5 g
AAB2085914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2085906 Supplier Thermo Scientific Chemicals Supplier No. B2085906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 597-76-2 | Molecular Formula: C8H18O | Molecular Weight: 130.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 597-76-2
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00039616
InChI Key WNDLTOTUHMHNOC-UHFFFAOYSA-N
Synonym 3-ethyl-3-hexanol, 3-hexanol, 3-ethyl, 1,1-diethyl-1-butanol, diethyl n-propyl carbinol, acmc-1ak6h
PubChem CID 69008
IUPAC Name 3-ethylhexan-3-ol
SMILES CCCC(O)(CC)CC

Specifications

Density 0.83
Quantity 5 g
Formula Weight 130.23
Percent Purity 98%
Chemical Name or Material 3-Ethyl-3-hexanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-908-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only