missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-2,4-dimethylpyrrole 97.0+%, TCI America™

Catalog No. E05555G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05555G 5 g
E05551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05555G Supplier TCI America Supplier No. E05555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 517-22-6
Molecular Formula C8H13N
Molecular Weight (g/mol) 123.199
MDL Number MFCD00005222
InChI Key ZEBBLOXDLGIMEG-UHFFFAOYSA-N
Synonym 2,4-dimethyl-3-ethylpyrrole, kryptopyrrole, kryptopyrrol, 3-ethyl-2,4-dimethylpyrrole, cryptopyrrole, 2,4-dimethyl-3-ethyl-1h-pyrrole, 1h-pyrrole, 3-ethyl-2,4-dimethyl, pyrrole, 3-ethyl-2,4-dimethyl, acmc-209kvh, 5-20-05-00099 beilstein handbook reference
PubChem CID 10600
IUPAC Name 3-ethyl-2,4-dimethyl-1H-pyrrole
SMILES CCC1=C(NC=C1C)C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 123.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Ethyl-2,4-dimethylpyrrole
RTECSNumber UX9629500
TSCA No
Recommended Storage Refrigerator