missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-1-pentyn-3-ol, 98%

Catalog No. AAL0633514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0633514 25 g
AAL0633506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0633514 Supplier Thermo Scientific Chemicals Supplier No. L0633514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6285-06-9 | C7H12O | 112.17 g/mol

3-Ethyl-1-pentyn-3-ol is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethyl-1-pentyn-3-ol is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.

Chemical Identifiers

CAS 6285-06-9
MDL Number MFCD00014392

Specifications

Density 0.873
Boiling Point 138°C to 140°C
Flash Point 38°C (100°F)
Molecular Formula C7H12O
Refractive Index 1.438
Quantity 25 g
UN Number UN1987
Beilstein 1740944
Synonym 3-ethyl-1-pentyn-3-ol, 1-pentyn-3-ol, 3-ethyl, diethylethynylcarbinol, 3-pentanol, 3-ethynyl, diaethyl-aethenyl-carbinol german, 3-aethyl-pentin-1-ol-3, 3-aethyl-pentin-1-ol-3 german, d iethylethynylcarbinol, acmc-1bagf, diaethyl-aethenyl-carbinol
Solubility Information Not miscible in water.
InChI Key PUNRPAWKFTXZIW-UHFFFAOYSA-N
SMILES CCC(CC)(C#C)O
IUPAC Name 3-ethylpent-1-yn-3-ol
Molecular Weight (g/mol) 112.17
PubChem CID 22698
Formula Weight 112.17
Percent Purity 98%
Chemical Name or Material 3-Ethyl-1-pentyn-3-ol
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 228-512-1
RTECSNumber SC4857000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only