missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-1-pentyn-3-ol 97.0+%, TCI America™

Catalog No. p-7128914
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E027325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E027325ML Supplier TCI America Supplier No. E027325ML

Chemical Identifiers

CAS 6285-06-9
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00014392
InChI Key PUNRPAWKFTXZIW-UHFFFAOYSA-N
Synonym 3-ethyl-1-pentyn-3-ol, 1-pentyn-3-ol, 3-ethyl, diethylethynylcarbinol, 3-pentanol, 3-ethynyl, diaethyl-aethenyl-carbinol german, 3-aethyl-pentin-1-ol-3, 3-aethyl-pentin-1-ol-3 german, d iethylethynylcarbinol, acmc-1bagf, diaethyl-aethenyl-carbinol
PubChem CID 22698
IUPAC Name 3-ethylpent-1-yn-3-ol
SMILES CCC(CC)(C#C)O

Specifications

Color Yellow
Boiling Point 138°C
Quantity 25 mL
UN Number 1987
Formula Weight 112.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Ethyl-1-pentyn-3-ol
RTECSNumber SC4857000
TSCA No