missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethoxypropionitrile 99.0+%, TCI America™
$52.19 - $143.54
Chemical Identifiers
| CAS | 2141-62-0 |
|---|---|
| Molecular Formula | C5H9NO |
| Molecular Weight (g/mol) | 99.133 |
| MDL Number | MFCD00001959 |
| InChI Key | DCWQZPJHHVLHSV-UHFFFAOYSA-N |
| Synonym | 3-ethoxypropionitrile, propanenitrile, 3-ethoxy, propionitrile, 3-ethoxy, 3-ethoxy-propionitrile, 3-ethoxypropiononitrile, beta-ethoxypropionitrile, 2-ethoxypropionitrile, alpha-ethoxypropionitrile, propionitrile, 2-ethoxy, .beta.-ethoxypropionitrile |
| PubChem CID | 16501 |
| IUPAC Name | 3-ethoxypropanenitrile |
| SMILES | CCOCCC#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E029925ML
|
TCI America
E029925ML |
25 mL |
Each for $52.19
|
|
|||||
|
E0299500ML
|
TCI America
E0299500ML |
500 mL |
Each for $143.54
|
|
|||||
Chemical Identifiers
| 2141-62-0 | |
| 99.133 | |
| DCWQZPJHHVLHSV-UHFFFAOYSA-N | |
| 16501 | |
| CCOCCC#N |
| C5H9NO | |
| MFCD00001959 | |
| 3-ethoxypropionitrile, propanenitrile, 3-ethoxy, propionitrile, 3-ethoxy, 3-ethoxy-propionitrile, 3-ethoxypropiononitrile, beta-ethoxypropionitrile, 2-ethoxypropionitrile, alpha-ethoxypropionitrile, propionitrile, 2-ethoxy, .beta.-ethoxypropionitrile | |
| 3-ethoxypropanenitrile |
Specifications
| 2141-62-0 | |
| 171°C | |
| MFCD00001959 | |
| 3-ethoxypropionitrile, propanenitrile, 3-ethoxy, propionitrile, 3-ethoxy, 3-ethoxy-propionitrile, 3-ethoxypropiononitrile, beta-ethoxypropionitrile, 2-ethoxypropionitrile, alpha-ethoxypropionitrile, propionitrile, 2-ethoxy, .beta.-ethoxypropionitrile | |
| CCOCCC#N | |
| 99.133 | |
| 99.13 | |
| Liquid |
| Yellow | |
| C5H9NO | |
| 25 mL | |
| DCWQZPJHHVLHSV-UHFFFAOYSA-N | |
| 3-ethoxypropanenitrile | |
| 16501 | |
| ≥99.0% (GC) | |
| 3-Ethoxypropionitrile |
Safety and Handling
EINECSNumber : (2)-1543
RTECSNumber : TZ4680000
TSCA : Yes