missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethoxypropionic Acid 98.0+%, TCI America™
$68.97 - $212.51
Chemical Identifiers
| CAS | 4324-38-3 |
|---|---|
| Molecular Formula | C5H10O3 |
| Molecular Weight (g/mol) | 118.132 |
| MDL Number | MFCD00058997 |
| InChI Key | JRXXEXVXTFEBIY-UHFFFAOYSA-N |
| Synonym | O-Ethylhydracrylic Acid |
| PubChem CID | 61351 |
| IUPAC Name | 3-ethoxypropanoic acid |
| SMILES | CCOCCC(=O)O |
Chemical Identifiers
| 4324-38-3 | |
| 118.132 | |
| JRXXEXVXTFEBIY-UHFFFAOYSA-N | |
| 61351 | |
| CCOCCC(=O)O |
| C5H10O3 | |
| MFCD00058997 | |
| O-Ethylhydracrylic Acid | |
| 3-ethoxypropanoic acid |
Specifications
| 4324-38-3 | |
| 219°C | |
| MFCD00058997 | |
| O-Ethylhydracrylic Acid | |
| CCOCCC(=O)O | |
| 118.132 | |
| 118.13 | |
| Liquid |
| Colorless | |
| C5H10O3 | |
| 5 mL | |
| JRXXEXVXTFEBIY-UHFFFAOYSA-N | |
| 3-ethoxypropanoic acid | |
| 61351 | |
| ≥98.0% (GC,T) | |
| 3-Ethoxypropionic Acid |
Safety and Handling
TSCA : No