missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxymethylene-2,4-pentanedione, 97+%

Catalog No. AAB2544506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2544506 5 g
AAB2544514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2544506 Supplier Thermo Scientific Chemicals Supplier No. B2544506

CAS: 33884-41-2 | C8H12O3 | 156.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33884-41-2
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.18
MDL Number MFCD07368652
InChI Key ITTXGKOHFZJUEX-UHFFFAOYSA-N
PubChem CID 240418
IUPAC Name 3-(ethoxymethylidene)pentane-2,4-dione
SMILES CCOC=C(C(C)=O)C(C)=O

Specifications

Boiling Point 140°C to 142°C (15 mmHg)
Refractive Index 1.484
Quantity 5 g
Synonym 3-ethoxymethylene pentane-2,4-dione, 3-ethoxymethylene-2,4-pentanedione, 3-ethoxymethylidene pentane-2,4-dione, acmc-1cmxl, 3-acetyl-4-ethoxy-3-buten-2-one, 3-ethoxymethylene-pentan-2,4-dione, 2,4-pentanedione,3-ethoxymethylene, 2,4-pentanedione, 3-ethoxymethylene, methyl 1-methylcarbonyl-2-ethoxyvinyl ketone
Formula Weight 156.18
Percent Purity ≥98%
Chemical Name or Material 3-Ethoxymethylene-2,4-pentanedione
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only