missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Ethoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$35.41 - $35.41
Chemical Identifiers
| CAS | 4334-87-6 |
|---|---|
| Molecular Formula | C9H11BO4 |
| Molecular Weight (g/mol) | 193.99 |
| MDL Number | MFCD02179444 |
| InChI Key | REHVCPNQQBDOJJ-UHFFFAOYSA-N |
| PubChem CID | 3249312 |
| IUPAC Name | [3-(ethoxycarbonyl)phenyl]boronic acid |
| SMILES | CCOC(=O)C1=CC=CC(=C1)B(O)O |
Chemical Identifiers
| 4334-87-6 | |
| 193.99 | |
| REHVCPNQQBDOJJ-UHFFFAOYSA-N | |
| [3-(ethoxycarbonyl)phenyl]boronic acid |
| C9H11BO4 | |
| MFCD02179444 | |
| 3249312 | |
| CCOC(=O)C1=CC=CC(=C1)B(O)O |
Specifications
| 4334-87-6 | |
| White | |
| MFCD02179444 | |
| 3-ethoxycarbonyl phenylboronic acid, 3-ethoxycarbonylphenyl boronic acid, 3-carbethoxyphenylboronic acid, m-ethoxycarbonylphenylboronic acid, 3-ethoxycarbonyl phenyl boronic acid, ethyl 3-dihydroxyboranyl benzoate, ethyl 3-boronobenzoate, 3-ethoxycarbonylbenzeneboronic acid, 3-ethoxycarbonyl benzeneboronic acid | |
| CCOC(=O)C1=CC=CC(=C1)B(O)O | |
| 193.99 | |
| 193.99 | |
| 3-(Ethoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride) |
| 138°C | |
| C9H11BO4 | |
| 1 g | |
| REHVCPNQQBDOJJ-UHFFFAOYSA-N | |
| [3-(ethoxycarbonyl)phenyl]boronic acid | |
| 3249312 | |
| Crystalline Powder |
Safety and Handling
TSCA : No