missing translation for 'onlineSavingsMsg'
Learn More

3-(Ethoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. E08481G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E08481G Supplier TCI America Supplier No. E08481G

Chemical Identifiers

CAS 4334-87-6
Molecular Formula C9H11BO4
Molecular Weight (g/mol) 193.99
MDL Number MFCD02179444
InChI Key REHVCPNQQBDOJJ-UHFFFAOYSA-N
PubChem CID 3249312
IUPAC Name [3-(ethoxycarbonyl)phenyl]boronic acid
SMILES CCOC(=O)C1=CC=CC(=C1)B(O)O

Specifications

Melting Point 138°C
Color White
Quantity 1 g
Synonym 3-ethoxycarbonyl phenylboronic acid, 3-ethoxycarbonylphenyl boronic acid, 3-carbethoxyphenylboronic acid, m-ethoxycarbonylphenylboronic acid, 3-ethoxycarbonyl phenyl boronic acid, ethyl 3-dihydroxyboranyl benzoate, ethyl 3-boronobenzoate, 3-ethoxycarbonylbenzeneboronic acid, 3-ethoxycarbonyl benzeneboronic acid
Formula Weight 193.99
Physical Form Crystalline Powder
Chemical Name or Material 3-(Ethoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride)