missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-2-nitropyridine 98.0+%, TCI America™

Catalog No. p-7128976
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03901G 1 g
E03905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E03901G Supplier TCI America Supplier No. E03901G

Chemical Identifiers

CAS 74037-50-6
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.15
MDL Number MFCD00006257
InChI Key WBEVIQNYYGPESH-UHFFFAOYSA-N
PubChem CID 52866
IUPAC Name 3-ethoxy-2-nitropyridine
SMILES CCOC1=CC=CN=C1[N+]([O-])=O

Specifications

Boiling Point 175°C
Quantity 1 g
Synonym pyridine, 3-ethoxy-2-nitro, 3-ethoxy-2-nitro-pyridine, 2-nitro-3-ethoxy pyridine, pubchem6624, acmc-209otd, 3-ethoxy-2-nitropyridin, 2-nitro-3-ethoxypyridine, pyridine,3-ethoxy-2-nitro, 4-21-00-00436 beilstein handbook reference
Formula Weight 168.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Ethoxy-2-nitropyridine
RTECSNumber UT3001500
TSCA No