missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-2-cyclohexen-1-one 98.0+%, TCI America™

Catalog No. p-7128974
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03845ML 5 mL
E038425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E03845ML Supplier TCI America Supplier No. E03845ML
Only null left

Chemical Identifiers

CAS 5323-87-5
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00001580
InChI Key JWCFJPLIRVYENQ-UHFFFAOYSA-N
Synonym 3-ethoxy-2-cyclohexen-1-one, 3-ethoxycyclohex-2-enone, 2-cyclohexen-1-one, 3-ethoxy, 3-ethoxy-2-cyclohexenone, 3-ethoxy-2-cyclohexene-1-one, 3-ethoxycyclohex-2-ene-1-one, 1-ethoxycyclohexene-3-one, acmc-209l4y, 3-ethoxy-2-cyclohexen-one, 3-ethoxy-cyclohex-2-enone
PubChem CID 79216
IUPAC Name 3-ethoxycyclohex-2-en-1-one
SMILES CCOC1=CC(=O)CCC1

Specifications

Color Yellow
Boiling Point 119°C
Quantity 5 mL
Formula Weight 140.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Ethoxy-2-cyclohexen-1-one