missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-2-cyclohexen-1-one, 99%

CAS: 5323-87-5 | C8H12O2 | 140.18 g/mol

Supplier:  Thermo Scientific Chemicals A1422418

 View more versions of this product

Catalog No. AAA1422418


Only null left
Explore available promotions

Description

Description

3-Ethoxy-2-cyclohexen-1-one is a useful intermediate for the preparation of 3-alkyl- or aryl-2-cyclohexen-1-ones, by addition of organometallic reagents and subsequent hydrolysis. It helps in kinetic enolate formation (LDA) and alkylation occurs at the 6-position, the Stork-Danheiser kinetic alkylation procedure, which has found extensive use in alicyclic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethoxy-2-cyclohexen-1-one is a useful intermediate for the preparation of 3-alkyl- or aryl-2-cyclohexen-1-ones, by addition of organometallic reagents and subsequent hydrolysis. It helps in kinetic enolate formation (LDA) and alkylation occurs at the 6-position, the Stork-Danheiser kinetic alkylation procedure, which has found extensive use in alicyclic synthesis.

Solubility
Slightly soluble in water.

Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents.
Specifications

Specifications

3-Ethoxy-2-cyclohexen-1-one
1.04
107°C (224°F)
1.506
50 g
3-ethoxy-2-cyclohexen-1-one, 3-ethoxycyclohex-2-enone, 2-cyclohexen-1-one, 3-ethoxy, 3-ethoxy-2-cyclohexenone, 3-ethoxy-2-cyclohexene-1-one, 3-ethoxycyclohex-2-ene-1-one, 1-ethoxycyclohexene-3-one, acmc-209l4y, 3-ethoxy-2-cyclohexen-one, 3-ethoxy-cyclohex-2-enone
JWCFJPLIRVYENQ-UHFFFAOYSA-N
3-ethoxycyclohex-2-en-1-one
79216
99%
5323-87-5
76°C to 78°C (1 mmHg)
C8H12O2
MFCD00001580
2042049
Slightly soluble in water.
CCOC1=CC(=O)CCC1
140.18
140.18

Chemical Identifiers

5323-87-5
140.18
JWCFJPLIRVYENQ-UHFFFAOYSA-N
79216
CCOC1=CC(=O)CCC1
C8H12O2
MFCD00001580
3-ethoxy-2-cyclohexen-1-one, 3-ethoxycyclohex-2-enone, 2-cyclohexen-1-one, 3-ethoxy, 3-ethoxy-2-cyclohexenone, 3-ethoxy-2-cyclohexene-1-one, 3-ethoxycyclohex-2-ene-1-one, 1-ethoxycyclohexene-3-one, acmc-209l4y, 3-ethoxy-2-cyclohexen-one, 3-ethoxy-cyclohex-2-enone
3-ethoxycyclohex-2-en-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 226-190-7

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only