missing translation for 'onlineSavingsMsg'
Learn More

3-(Dodecylthio)propionic Acid 90.0+%, TCI America™

Catalog No. p-7127933
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D381425G 25 g
D3814500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D381425G Supplier TCI America Supplier No. D381425G

Chemical Identifiers

CAS 1462-52-8
Molecular Formula C15H30O2S
Molecular Weight (g/mol) 274.463
InChI Key VKLOPQHLJNFYKK-UHFFFAOYSA-N
Synonym 3-(Laurylthio)propionic Acid
PubChem CID 73834
IUPAC Name 3-dodecylsulfanylpropanoic acid
SMILES CCCCCCCCCCCCSCCC(=O)O

Specifications

Melting Point 61°C
Color White-Yellow
Quantity 25 g
Formula Weight 274.46
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Dodecylthio)propionic Acid
EINECSNumber (2)-1253&(2)-1388
TSCA Yes