missing translation for 'onlineSavingsMsg'
Learn More

3-Dodecyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene 95.0+%, TCI America™

Catalog No. D42901G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42901G 1 g
D42905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42901G Supplier TCI America Supplier No. D42901G
Only null left

Chemical Identifiers

CAS 960524-18-9
Molecular Formula C22H39BO2S
Molecular Weight (g/mol) 378.422
InChI Key VOFQGWRXFFSUGR-UHFFFAOYSA-N
Synonym 2-(3-Dodecyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Dodecyl-2-thiopheneboronic Acid Pinacol Ester
PubChem CID 58409892
IUPAC Name 2-(3-dodecylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=CS2)CCCCCCCCCCCC

Specifications

Color Yellow
Quantity 1 g
Formula Weight 378.42
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Dodecyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene
TSCA No
Recommended Storage Refrigerator