Learn More
3-Dimethylaminopropylchloride hydrochloride, 99%
CAS: 5407-04-5 | C5H13Cl2N | 158.07 g/mol
$218.63 - $218.63
Chemical Identifiers
| CAS | 4-5-5407 |
|---|---|
| Molecular Weight (g/mol) | 158.07 |
| MDL Number | MFCD00012521 |
| InChI Key | LJQNMDZRCXJETK-UHFFFAOYSA-N |
| Synonym | 3-dimethylaminopropylchloride hydrochloride, 3-chloro-n,n-dimethylpropan-1-amine hydrochloride, 3-dimethylaminopropyl chloride hydrochloride, 3-dimethylamino propyl chloride hydrochloride, n,n-dimethyl-3-chloropropylamine hydrochloride, 3-chloropropyl dimethylamine hydrochloride, 3-chloro-n,n-dimethylpropylamine hydrochloride, dimethylaminopropyl chloride hydrochloride, ccris 7054, dimethylaminopropyl chloride, hydrochloride |
| PubChem CID | 94308 |
| IUPAC Name | 3-chloro-N,N-dimethylpropan-1-amine;hydrochloride |
| SMILES | CN(C)CCCCl.Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC115915000
|
Thermo Scientific Chemicals
115915000 |
500 g | Glass bottle |
Each for $218.63
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4-5-5407 | |
| MFCD00012521 | |
| 3-dimethylaminopropylchloride hydrochloride, 3-chloro-n,n-dimethylpropan-1-amine hydrochloride, 3-dimethylaminopropyl chloride hydrochloride, 3-dimethylamino propyl chloride hydrochloride, n,n-dimethyl-3-chloropropylamine hydrochloride, 3-chloropropyl dimethylamine hydrochloride, 3-chloro-n,n-dimethylpropylamine hydrochloride, dimethylaminopropyl chloride hydrochloride, ccris 7054, dimethylaminopropyl chloride, hydrochloride | |
| 3-chloro-N,N-dimethylpropan-1-amine;hydrochloride |
| 158.07 | |
| LJQNMDZRCXJETK-UHFFFAOYSA-N | |
| 94308 | |
| CN(C)CCCCl.Cl |
Specifications
| 4-5-5407 | |
| 100.0 | |
| White to Yellow | |
| Authentic | |
| Glass bottle | |
| (CH3)2N(CH2)3Cl·HCl | |
| 500 g | |
| 3-dimethylaminopropylchloride hydrochloride, 3-chloro-n,n-dimethylpropan-1-amine hydrochloride, 3-dimethylaminopropyl chloride hydrochloride, 3-dimethylamino propyl chloride hydrochloride, n,n-dimethyl-3-chloropropylamine hydrochloride, 3-chloropropyl dimethylamine hydrochloride, 3-chloro-n,n-dimethylpropylamine hydrochloride, dimethylaminopropyl chloride hydrochloride, ccris 7054, dimethylaminopropyl chloride, hydrochloride | |
| LJQNMDZRCXJETK-UHFFFAOYSA-N | |
| 3-chloro-N,N-dimethylpropan-1-amine;hydrochloride | |
| 94308 | |
| 99% | |
| 3-Dimethylaminopropylchloride hydrochloride, 99% |
| 98.5 | |
| 139.0°C to 145.0°C | |
| 1% max. (105°C, 3 hrs) | |
| 99% | |
| C5H12ClN·HCl | |
| MFCD00012521 | |
| 04, 148 | |
| Solubility in water: soluble 2000g/L in water (20°C). Other solubilities: soluble in ethanol, very soluble in methanol, insoluble in toluene and acetone | |
| CN(C)CCCCl.Cl | |
| 158.07 | |
| 158.07 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 226-467-2
RTECSNumber : UI0209950
TSCA : TSCA
RUO – Research Use Only