missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylaminopropyl chloride, 97%, stab. with 0.5% oxalic acid

Catalog No. p-7056452
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6537103 1 g
AAH6537106 5 g
AAH6537114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6537103 Supplier Thermo Scientific Chemicals Supplier No. H6537103
Only null left

CAS: 109-54-6 | C5H12ClN | 121.608 g/mol

It is used in chlorpromazine, anti-inflammatory pain static, propylene clomipramine work, doxpin and amy for butyl intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in chlorpromazine, anti-inflammatory pain static, propylene clomipramine work, doxpin and amy for butyl intermediates.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 109-54-6
Molecular Formula C5H12ClN
Molecular Weight (g/mol) 121.608
MDL Number MFCD00044496
InChI Key NYYRRBOMNHUCLB-UHFFFAOYSA-N
Synonym 3-chloro-1-n,n-dimethyl propylamine, dimethylaminopropyl chloride, 3-chloropropyl dimethylamine, 3-chloropropyl dimethyl amine, 1-dimethylamino-3-chloropropane, 1-propanamine, 3-chloro-n,n-dimethyl, n-3-chloropropyl dimethylamine, 3-chloro-propyl-dimethyl-amine, 3-dimethylaminopropyl chloride, 3-dimethylaminopropylchloride
PubChem CID 66960
IUPAC Name 3-chloro-N,N-dimethylpropan-1-amine
SMILES CN(C)CCCCl

Specifications

Melting Point 168°C
Color Colorless
Density 0.929 g/mL
Refractive Index 1.432
Linear Formula Cl(CH2)3N(CH3)2
Quantity 1 g
Beilstein 605294
Sensitivity Air sensitive
Solubility Information Slightly soluble in water.
Formula Weight 121.61
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Dimethylaminopropyl chloride
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 203-679-3
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only