missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylaminomethyl)-5-nitroindole, 95%, Thermo Scientific Chemicals

Catalog No. AAH6393903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6393903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6393903 Supplier Thermo Scientific Chemicals Supplier No. H6393903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3414-64-0 | C11H13N3O2 | 219.244 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3414-64-0
Molecular Formula C11H13N3O2
Molecular Weight (g/mol) 219.244
MDL Number MFCD00544197
InChI Key SVNVKGIWGOHXPW-UHFFFAOYSA-N
Synonym 5-nitrogramine, n,n-dimethyl-1-5-nitro-1h-indol-3-yl methanamine, 1h-indole-3-methanamine, n,n-dimethyl-5-nitro, indole, 3-dimethylamino methyl-5-nitro, 3-dimethylamino methyl-5-nitroindole, dimethyl 5-nitro-1h-indol-3-yl methyl amine, 4-22-00-04317 beilstein handbook reference, 3-dimethylaminomethyl-5-nitroindole, 1h-indole-3-methanamine,n-dimethyl-5-nitro
PubChem CID 18882
IUPAC Name N,N-dimethyl-1-(5-nitro-1H-indol-3-yl)methanamine
SMILES CN(C)CC1=CNC2=C1C=C(C=C2)[N+](=O)[O-]

Specifications

Quantity 1 g
Beilstein 216692
Formula Weight 219.24
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(Dimethylaminomethyl)-5-nitroindole
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber NL7710000
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only