missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylamino)pyrrolidine 98.0+%, TCI America™

Catalog No. D185925G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D185925G 25 g
D185910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D185925G Supplier TCI America Supplier No. D185925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 69478-75-7
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD01569246
InChI Key AVAWMINJNRAQFS-UHFFFAOYSA-N
PubChem CID 2758519
IUPAC Name N,N-dimethylpyrrolidin-3-amine
SMILES CN(C)C1CCNC1

Specifications

Color Yellow
Boiling Point 164°C
Quantity 25 g
UN Number 2734
Formula Weight 114.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Dimethylamino)pyrrolidine