missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylamino)phenylboronic Acid Hydrochloride (contains varying amounts of Anhydride), TCI America™

Catalog No. D40131G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D40131G Supplier TCI America Supplier No. D40131G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3-(Dimethylamino)phenylboronic Acid Hydrochloride (contains varying amounts of Anhydride)
CAS 1256355-23-3
Color White
Molecular Formula C8H13BClNO2
Quantity 1 g
MDL Number MFCD04112542
Synonym 3-(Dimethylamino)benzeneboronic Acid Hydrochloride
InChI Key QDYZZIRCCVRLBI-UHFFFAOYSA-N
SMILES Cl.CN(C)C1=CC=CC(=C1)B(O)O
IUPAC Name [3-(dimethylamino)phenyl]boronic acid hydrochloride
Molecular Weight (g/mol) 201.46
PubChem CID 44754897
Formula Weight 201.46
Percent Purity 98 to 110%
Physical Form Crystalline Powder
Show More Show Less