missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylamino)phenol 97.0+%, TCI America™

Catalog No. D065725G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D065725G 25 g
D0657100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D065725G Supplier TCI America Supplier No. D065725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99-07-0
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00002264
InChI Key MESJRHHDBDCQTH-UHFFFAOYSA-N
Synonym 3-dimethylamino phenol, phenol, 3-dimethylamino, m-dimethylamino phenol, m-dimethylaminophenol, n,n-dimethyl-m-aminophenol, n,n-dimethyl-3-aminophenol, phenol, m-dimethylamino, 3-hydroxyphenyl dimethylamine, 3-hydroxy-n,n-dimethylaniline, 3-n,n-dimethylaminophenol
PubChem CID 7421
IUPAC Name 3-(dimethylamino)phenol
SMILES CN(C)C1=CC=CC(O)=C1

Specifications

Melting Point 86°C
Color Gray-Red
Boiling Point 268°C
Quantity 25 g
Formula Weight 137.18
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Dimethylamino)phenol
EINECSNumber (9)-2069
RTECSNumber SL1050000
TSCA Yes