missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylamino)acrolein 98.0+%, TCI America™

Catalog No. p-7128094
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40925G 5 g
D409225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40925G Supplier TCI America Supplier No. D40925G

Chemical Identifiers

CAS 927-63-9
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00006999
InChI Key RRLMPLDPCKRASL-ONEGZZNKSA-N
Synonym 3-dimethylaminoacrolein, 3-dimethylaminoacrylaldehyde, 3-dimethylamino acrylaldehyde, 3-dimethylamino acrolein, 2-propenal, 3-dimethylamino, e-3-dimethylamino acrylaldehyde, e-3-dimethylamino prop-2-enal, 3-dimethylamino propenal, 2e-3-dimethylamino prop-2-enal, .beta.-dimethylamino acrolein
PubChem CID 638320
IUPAC Name (2E)-3-(dimethylamino)prop-2-enal
SMILES CN(C)\C=C\C=O

Specifications

Boiling Point 91°C
Quantity 5 g
UN Number 3267
Formula Weight 99.13
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Dimethylamino)acrolein