missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Dimethylamino)acrolein 98.0+%, TCI America™
Supplier: TCI America D409225G
Specifications
| 3-(Dimethylamino)acrolein | |
| 91°C | |
| MFCD00006999 | |
| 3267 | |
| RRLMPLDPCKRASL-ONEGZZNKSA-N | |
| (2E)-3-(dimethylamino)prop-2-enal | |
| 638320 | |
| ≥98.0% (GC,T) |
| 927-63-9 | |
| C5H9NO | |
| 25 g | |
| 3-dimethylaminoacrolein, 3-dimethylaminoacrylaldehyde, 3-dimethylamino acrylaldehyde, 3-dimethylamino acrolein, 2-propenal, 3-dimethylamino, e-3-dimethylamino acrylaldehyde, e-3-dimethylamino prop-2-enal, 3-dimethylamino propenal, 2e-3-dimethylamino prop-2-enal, .beta.-dimethylamino acrolein | |
| CN(C)\C=C\C=O | |
| 99.13 | |
| 99.13 | |
| Liquid |
Chemical Identifiers
| 927-63-9 | |
| 99.13 | |
| RRLMPLDPCKRASL-ONEGZZNKSA-N | |
| 638320 | |
| CN(C)\C=C\C=O |
| C5H9NO | |
| MFCD00006999 | |
| 3-dimethylaminoacrolein, 3-dimethylaminoacrylaldehyde, 3-dimethylamino acrylaldehyde, 3-dimethylamino acrolein, 2-propenal, 3-dimethylamino, e-3-dimethylamino acrylaldehyde, e-3-dimethylamino prop-2-enal, 3-dimethylamino propenal, 2e-3-dimethylamino prop-2-enal, .beta.-dimethylamino acrolein | |
| (2E)-3-(dimethylamino)prop-2-enal |
Safety and Handling
TSCA : No