missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylamino-2-methyl-2-propenal 97.0+%, TCI America™

Catalog No. d24835g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24835G 5 g
D248325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24835G Supplier TCI America Supplier No. D24835G

Chemical Identifiers

CAS 19125-76-9
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00075471
InChI Key JGAMOQYFDMQPRJ-XQRVVYSFSA-N
Synonym z-3-dimethylamino-2-methylacrylaldehyde, 2z-3-dimethylamino-2-methylacrylaldehyde, 2z-3-dimethylamino-2-methylprop-2-enal, z-3-dimethylamino-2-methylprop-2-enal, 2-propenal, 3-dimethylamino-2-methyl-, z, 2-propenal, 3-dimethylamino-2-methyl-, 2z
PubChem CID 5919306
IUPAC Name (Z)-3-(dimethylamino)-2-methylprop-2-enal
SMILES CC(=CN(C)C)C=O

Specifications

Quantity 5 g
Formula Weight 113.16
Percent Purity ≥97.0% (GC)
Physical Form Crystals
Chemical Name or Material 3-Dimethylamino-2-methyl-2-propenal