missing translation for 'onlineSavingsMsg'
Learn More

3-(Difluoromethyl)pyridine, 97%

Catalog No. p-7031919
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33189MD 250 mg
AAH3318903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33189MD Supplier Thermo Scientific Chemicals Supplier No. H33189MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 76541-44-1 | C6H5F2N | 129.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 76541-44-1
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
MDL Number MFCD11226584
InChI Key HKRKGTFBASUFRO-UHFFFAOYSA-N
Synonym 3-difluoromethyl pyridine, pyridine, 3-difluoromethyl, pyridine,3-difluoromethyl, pyridine, 3-difluoromethyl-9ci
PubChem CID 640475
IUPAC Name 3-(difluoromethyl)pyridine
SMILES FC(F)C1=CC=CN=C1

Specifications

Boiling Point 42°C (6 mmHg)
Flash Point 54°C (129°F)
Quantity 250 mg
UN Number UN1993
Formula Weight 129.11
Percent Purity 97%
Chemical Name or Material 3-(Difluoromethyl)pyridine
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 278-487-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only