missing translation for 'onlineSavingsMsg'
Learn More

3-(Difluoromethyl)aniline, 97%

Catalog No. p-7032643
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34188MD 250 mg
AAH3418803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH34188MD Supplier Thermo Scientific Chemicals Supplier No. H34188MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 368-99-0 | C7H7F2N | 143.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 368-99-0
Molecular Formula C7H7F2N
Molecular Weight (g/mol) 143.14
MDL Number MFCD18384830
InChI Key IDFPXKDJNDYDKA-UHFFFAOYSA-N
Synonym 3-difluoromethyl aniline, 3-difluoromethyl benzenamine, 3-difluoromethyl-phenylamine, 3-difluoromethyl benzenamine hcl
PubChem CID 23445119
IUPAC Name 3-(difluoromethyl)aniline
SMILES NC1=CC=CC(=C1)C(F)F

Specifications

Boiling Point 82°C (4 mmHg)
Quantity 250 mg
UN Number UN2810
Formula Weight 143.14
Percent Purity 97%
Chemical Name or Material 3-(Difluoromethyl)aniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only